siRNA Details
| Gene Symbol: Ubqln4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033526.2 | NP_277068.1 | 6 | 1038 | ||
| GenBank mRNA | AB040050.1 | - | 6 | 1038 | ||
| AK150211.1 | - | 6 | 976 | |||
| AK156515.1 | - | 6 | 992 | |||
| AK160578.1 | - | 6 | 990 | |||
| BC017686.1 | - | 6 | 1000 | 4236887 | ||

Ubqln4
Genomic Map