siRNA Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025772.4 | NP_080048.2 | 3 | 261 | ||
| GenBank mRNA | AJ404859.1 | - | 3 | 260 | ||
| AK150306.1 | - | 3 | 257 | |||
| AK159656.1 | - | 3 | 386 | |||
| BC018350.1 | - | 3 | 252 | 4195234 | ||
| BC048682.1 | - | 3 | 306 | |||
| BC058574.1 | - | 3 | 234 | 6487720 | ||

Dtnbp1
Genomic Map