siRNA Details
| Gene Symbol: Tmem40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168256.1 | NP_001161728.1 | 10 | 774 | ||
| NM_001168257.1 | NP_001161729.1 | 9 | 699 | |||
| NM_001168258.1 | NP_001161730.1 | 5 | 499 | |||
| NM_001168259.1 | NP_001161731.1 | 6 | 266 | |||
| NM_144805.2 | NP_659054.2 | 10 | 642 | |||
| GenBank mRNA | AK078864.1 | - | 6 | 267 | ||
| BC019416.1 | - | 10 | 604 | 5151159 | ||
| BX511925.1 | - | 5 | 226 | |||
| CT010303.1 | - | 9 | 572 | |||
| CV561368.1 | - | 9 | 641 | |||
| CX564997.1 | - | 10 | 673 | |||

Tmem40
Genomic Map