siRNA Details
| Gene Symbol: Hmgn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026122.4 | NP_080398.1 | 3, 4 | 253 | ||
| NM_175074.2 | NP_778249.1 | 3, 4 | 253 | |||
| GenBank mRNA | AK002970.1 | - | 3, 4 | 172 | ||
| AK003272.1 | - | 3, 4 | 201 | |||
| AK153223.1 | - | 3, 4 | 213 | |||
| BC005693.1 | - | 3, 4 | 189 | 3597594 | ||
| CD742335.1 | - | 3, 4 | 626 | |||

Hmgn3
Genomic Map