siRNA Details
| Gene Symbol: C77080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033189.3 | NP_001028361.1 | 6 | 2129 | ||
| NM_001285865.1 | NP_001272794.1 | 6 | 2019 | |||
| NM_001285866.1 | NP_001272795.1 | 6 | 1991 | |||
| NM_001285867.1 | NP_001272796.1 | 6 | 1880 | |||
| GenBank mRNA | AK038020.1 | - | 6 | 1970 | ||
| AK129382.1 | - | 6 | 1967 | |||
| AK154456.1 | - | 6 | 1881 | |||
| AK154932.1 | - | 6 | 2130 | |||

C77080
Genomic Map