siRNA Details
| Gene Symbol: A430005L14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163019.1 | NP_001156491.1 | 2 | 511 | ||
| NM_175287.4 | NP_780496.2 | 3 | 625 | |||
| GenBank mRNA | AK039812.1 | - | 3 | 626 | ||
| BC052148.1 | - | 3 | 625 | 6706797 | ||
| BE994111.1 | - | 2 | 481 | |||

A430005L14Rik
Genomic Map