siRNA Details
| Gene Symbol: C86187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135809.1 | - | 2 | 1264 | ||
| AK135845.1 | - | 4 | 1630 | |||
| AK135966.1 | - | 2 | 1269 | |||
| AK139730.1 | - | 2 | 1269 | |||
| AK163234.1 | - | 2 | 1266 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C86187| Gene Symbol: C86187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135809.1 | - | 2 | 1264 | ||
| AK135845.1 | - | 4 | 1630 | |||
| AK135966.1 | - | 2 | 1269 | |||
| AK139730.1 | - | 2 | 1269 | |||
| AK163234.1 | - | 2 | 1266 | |||