siRNA Details
Gene Symbol: C86187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135809.1 | - | 2 | 1298 | ||
AK135845.1 | - | 4 | 1664 | |||
AK135966.1 | - | 2 | 1303 | |||
AK163234.1 | - | 2 | 1300 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C86187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135809.1 | - | 2 | 1298 | ||
AK135845.1 | - | 4 | 1664 | |||
AK135966.1 | - | 2 | 1303 | |||
AK163234.1 | - | 2 | 1300 |