siRNA Details
| Gene Symbol: C86695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081662.1 | NP_001075131.1 | 5 | 620 | ||
| GenBank mRNA | AK135805.1 | - | 5 | 619 | ||
| DV648735.1 | - | 5 | 520 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C86695| Gene Symbol: C86695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081662.1 | NP_001075131.1 | 5 | 620 | ||
| GenBank mRNA | AK135805.1 | - | 5 | 619 | ||
| DV648735.1 | - | 5 | 520 | |||