siRNA Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153555.2 | NP_705783.1 | 4 | 607 | ||
| GenBank mRNA | AK035701.1 | - | 4 | 506 | ||
| AK047316.1 | - | 4 | 511 | |||
| AK146825.1 | - | 4 | 534 | |||
| AK162622.1 | - | 4 | 629 | |||
| AK164799.1 | - | 4 | 534 | |||
| AK167059.1 | - | 4 | 483 | |||
| AK217466.1 | - | 1 | 184 | |||

Dcaf8
Genomic Map