siRNA Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153555.2 | NP_705783.1 | 4 | 762 | ||
| GenBank mRNA | AK035701.1 | - | 4 | 661 | ||
| AK047316.1 | - | 4 | 666 | |||
| AK098114.1 | - | 4 | 781 | |||
| AK146825.1 | - | 4 | 689 | |||
| AK162622.1 | - | 4 | 784 | |||
| AK164799.1 | - | 4 | 689 | |||
| AK167059.1 | - | 4 | 638 | |||
| BC023704.1 | - | 5 | 703 | 5345701 | ||
| BC023804.1 | - | 5 | 816 | 5352941 | ||
| BC078641.1 | - | 4 | 1674 | |||

Dcaf8
Genomic Map