siRNA Details
| Gene Symbol: Lrrc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133807.1 | NP_598568.1 | 1, 2 | 291 | ||
| GenBank mRNA | AK053450.1 | - | 1, 2 | 318 | ||
| AK159374.1 | - | 1, 2 | 347 | |||
| AK159552.1 | - | 1, 2 | 341 | |||
| AK166576.1 | - | 1, 2 | 340 | |||
| AK167604.1 | - | 1, 2 | 282 | |||
| AK169250.1 | - | 1, 2 | 266 | |||
| AK169390.1 | - | 1, 2 | 290 | |||
| AK183165.1 | - | 1, 2 | 77 | |||
| BC006877.1 | - | 1, 2 | 291 | 2647513 | ||
| BC066172.1 | - | 1, 2 | 285 | 30429192 | ||

Lrrc59
Genomic Map