siRNA Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290699.1 | NP_001277628.1 | 8 | 1231 | ||
| NM_133817.3 | NP_598578.1 | 10 | 1469 | |||
| GenBank mRNA | AK030088.1 | - | 10 | 1302 | ||
| BC062154.1 | - | 8 | 1021 | 6331597 | ||

Zfp451
Genomic Map