siRNA Details
Gene Symbol: AI597479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133818.1 | NP_598579.1 | 2 | 243 | ||
GenBank mRNA | AK081372.1 | - | 2 | 234 | ||
BC006931.1 | - | 2 | 243 | 3598975 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AI597479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133818.1 | NP_598579.1 | 2 | 243 | ||
GenBank mRNA | AK081372.1 | - | 2 | 234 | ||
BC006931.1 | - | 2 | 243 | 3598975 |