siRNA Details
| Gene Symbol: Gtf3c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033194.2 | NP_001028366.1 | 5 | 708 | ||
| GenBank mRNA | AK145040.1 | - | 709 | |||
| AK160997.1 | - | 5 | 696 | |||
| AK165830.1 | - | 5 | 709 | |||
| BC141157.1 | - | 5 | 698 | 9055769 | ||

Gtf3c3
Genomic Map