siRNA Details
| Gene Symbol: Khdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113187.1 | NP_001106658.1 | 1, 2 | 475 | ||
| GenBank mRNA | AK139842.1 | - | 1, 2 | 475 | ||
| BG081161.2 | - | 1, 2 | 201 | |||
| Gene Symbol: Khdc1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC141037.1 | - | 115 | 9055649 | ||

Khdc1b
Genomic Map