siRNA Details
| Gene Symbol: Obsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178884.5 | NP_849215.3 | 2 | 1303 | ||
| GenBank mRNA | AK029863.1 | - | 2 | 1093 | ||
| AK031074.1 | - | 2 | 1304 | |||
| AK034852.1 | - | 2 | 1214 | |||
| AK048144.1 | - | 2 | 494 | |||
| AK162473.1 | - | 2 | 1213 | |||
| AK210724.1 | - | 1 | 71 | |||

Obsl1
Genomic Map