siRNA Details
| Gene Symbol: Myl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172118.1 | NP_742116.1 | 4 | 465 | ||
| GenBank mRNA | AK007972.1 | - | 4 | 475 | ||
| AK020258.1 | - | 4 | 466 | |||
| BC049974.1 | - | 4 | 500 | 4912899 | ||
| BC055439.1 | - | 4 | 489 | 2644903 | ||

Myl9
Genomic Map