siRNA Details
| Gene Symbol: Mrps26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207207.1 | NP_997090.1 | 1 | 177 | ||
| GenBank mRNA | AK131626.1 | - | 1 | 187 | ||
| AK169837.1 | - | 1 | 190 | |||
| BC048517.1 | - | 1 | 177 | 6706680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps26| Gene Symbol: Mrps26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207207.1 | NP_997090.1 | 1 | 177 | ||
| GenBank mRNA | AK131626.1 | - | 1 | 187 | ||
| AK169837.1 | - | 1 | 190 | |||
| BC048517.1 | - | 1 | 177 | 6706680 | ||