siRNA Details
| Gene Symbol: Ssx2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253768.1 | NP_001240697.1 | 4 | 722 | ||
| NM_001253769.1 | NP_001240698.1 | 3 | 618 | |||
| NM_001253770.1 | NP_001240699.1 | 3 | 467 | |||
| NM_138744.3 | NP_620083.1 | 3 | 467 | |||
| GenBank mRNA | AF532969.1 | - | 2 | 262 | ||
| AK031356.1 | - | 4 | 446 | |||
| AK043865.1 | - | 3 | 571 | |||
| AK049080.1 | - | 3 | 619 | |||
| AK077112.1 | - | 4 | 507 | |||
| AK173064.1 | - | 3 | 454 | |||
| BC021749.1 | - | 3 | 422 | 4165430 | ||
| BC031527.1 | - | 3 | 430 | 4008624 | ||

Ssx2ip
Genomic Map