siRNA Details
| Gene Symbol: Zscan29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290819.1 | NP_001277748.1 | 4 | 2295 | ||
| GenBank mRNA | CV555944.1 | - | 2 | 526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zscan29| Gene Symbol: Zscan29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290819.1 | NP_001277748.1 | 4 | 2295 | ||
| GenBank mRNA | CV555944.1 | - | 2 | 526 | ||