siRNA Details
| Gene Symbol: Dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026992.3 | NP_081268.1 | 5 | 921 | ||
| GenBank mRNA | AK006521.1 | - | 5 | 922 | ||
| AK011571.1 | - | 4 | 496 | |||
| BC030072.1 | - | 5 | 629 | 3376358 | ||
| BC091774.1 | - | 5 | 487 | 5150845 | ||
| Gene Symbol: Immp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012181.1 | - | 723 | |||

Dnajc24
Genomic Map