siRNA Details
| Gene Symbol: Abtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178890.3 | NP_849221.2 | 5 | 1855 | ||
| GenBank mRNA | AK042322.1 | - | 5 | 1853 | ||
| AK151620.1 | - | 5 | 1851 | |||
| AK152163.1 | - | 5 | 1851 | |||
| AK170569.1 | - | 5 | 1854 | |||
| AK172414.1 | - | 5 | 1853 | |||
| AK172468.1 | - | 5 | 1852 | |||
| BC054399.1 | - | 5 | 1855 | 6493840 | ||

Abtb2
Genomic Map