siRNA Details
| Gene Symbol: Golga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080968.1 | NP_001074437.1 | 22 | 2349 | ||
| NM_133852.2 | NP_598613.2 | 23 | 2430 | |||
| GenBank mRNA | AK076370.1 | - | 17 | 2048 | ||
| AK077516.1 | - | 21 | 2369 | |||
| AK220509.1 | - | 23 | 2358 | |||
| BC011407.1 | - | 21 | 2150 | 3596079 | ||

Golga2
Genomic Map