siRNA Details
| Gene Symbol: Tmem56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178936.3 | NP_849267.1 | 6 | 671 | ||
| GenBank mRNA | AK016295.1 | - | 8 | 925 | ||
| AK147601.1 | - | 6 | 719 | |||
| AK149544.1 | - | 6 | 696 | |||
| AK163285.1 | - | 6 | 709 | |||
| BC040365.1 | - | 6 | 671 | 4223855 | ||

Tmem56
Genomic Map