siRNA Details
Gene Symbol: Gucy2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130737.4 | NP_570093.1 | 2 | 229 | ||
XM_006229710.1 | XP_006229772.1 | 2 | 228 | |||
GenBank mRNA | L37203.1 | - | 2 | 229 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gucy2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130737.4 | NP_570093.1 | 2 | 229 | ||
XM_006229710.1 | XP_006229772.1 | 2 | 228 | |||
GenBank mRNA | L37203.1 | - | 2 | 229 |