siRNA Details
Gene Symbol: Bbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053618.1 | NP_446070.1 | 7 | 833 | ||
XM_006255131.2 | XP_006255193.1 | 7 | 992 | |||
XM_008772263.1 | XP_008770485.1 | 7 | 760 | |||
GenBank mRNA | AF342738.1 | - | 7 | 833 | ||
FQ222286.1 | - | 2 | 50 |
