siRNA Details
| Gene Symbol: Acsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053623.1 | NP_446075.1 | 13 | 1624 | ||
| XM_006257314.2 | XP_006257376.1 | 13 | 1962 | |||
| XM_006257315.2 | XP_006257377.1 | 14 | 1743 | |||
| XM_006257316.2 | XP_006257378.1 | 13 | 1767 | |||
| XM_008773385.1 | XP_008771607.1 | 14 | 2036 | |||
| GenBank mRNA | D85189.1 | - | 13 | 1624 | ||

Acsl4
Genomic Map