siRNA Details
| Gene Symbol: Kcnh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053630.1 | NP_446082.1 | 4 | 620 | ||
| GenBank mRNA | AB022699.1 | - | 4 | 620 | ||
| AJ007628.1 | - | 4 | 583 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnh4| Gene Symbol: Kcnh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053630.1 | NP_446082.1 | 4 | 620 | ||
| GenBank mRNA | AB022699.1 | - | 4 | 620 | ||
| AJ007628.1 | - | 4 | 583 | |||