siRNA Details
| Gene Symbol: St14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053635.2 | NP_446087.1 | 14, 15 | 1806 | ||
| GenBank mRNA | AB037898.1 | - | 14, 15 | 1680 | ||
| AB049189.1 | - | 14, 15 | 1806 | |||
| BC097271.1 | - | 14, 15 | 1711 | 7445508 | ||

St14
Genomic Map