siRNA Details
| Gene Symbol: Stxbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053637.2 | NP_446089.1 | 15 | 1454 | ||
| GenBank mRNA | AB046544.1 | - | 15 | 1455 | ||
| BC098660.1 | - | 15 | 1382 | 7386801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stxbp3| Gene Symbol: Stxbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053637.2 | NP_446089.1 | 15 | 1454 | ||
| GenBank mRNA | AB046544.1 | - | 15 | 1455 | ||
| BC098660.1 | - | 15 | 1382 | 7386801 | ||