siRNA Details
| Gene Symbol: Idh3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053638.1 | NP_446090.1 | 6 | 607 | ||
| GenBank mRNA | AB047541.1 | - | 6 | 607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Idh3a| Gene Symbol: Idh3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053638.1 | NP_446090.1 | 6 | 607 | ||
| GenBank mRNA | AB047541.1 | - | 6 | 607 | ||