siRNA Details
| Gene Symbol: Cdh23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053644.1 | NP_446096.1 | 31 | 4352 | ||
| GenBank mRNA | AB053447.1 | - | 31 | 4352 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh23| Gene Symbol: Cdh23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053644.1 | NP_446096.1 | 31 | 4352 | ||
| GenBank mRNA | AB053447.1 | - | 31 | 4352 | ||