siRNA Details
Gene Symbol: Bco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053648.3 | NP_446100.2 | 7 | 1029 | ||
GenBank mRNA | FQ219222.1 | - | 7 | 1080 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053648.3 | NP_446100.2 | 7 | 1029 | ||
GenBank mRNA | FQ219222.1 | - | 7 | 1080 |