siRNA Details
| Gene Symbol: Pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053650.1 | NP_446102.1 | 7 | 901 | ||
| GenBank mRNA | AF002281.1 | - | 7 | 919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdlim3| Gene Symbol: Pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053650.1 | NP_446102.1 | 7 | 901 | ||
| GenBank mRNA | AF002281.1 | - | 7 | 919 | ||