siRNA Details
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053657.1 | NP_446109.1 | 8 | 1592 | ||
GenBank mRNA | AF021935.1 | - | 8 | 1592 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053657.1 | NP_446109.1 | 8 | 1592 | ||
GenBank mRNA | AF021935.1 | - | 8 | 1592 |