siRNA Details
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053662.3 | NP_446114.1 | 602 | |||
XM_008761029.1 | XP_008759251.1 | 1678 | ||||
GenBank mRNA | AF030091.3 | - | 602 | |||
BC085686.1 | - | 656 | ||||
BC128699.1 | - | 696 | ||||
BC161817.1 | - | 548 | 9022416 | |||
FQ213168.1 | - | 1 | 26 |
