siRNA Details
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053662.3 | NP_446114.1 | 1351 | |||
XM_008761029.1 | XP_008759251.1 | 2427 | ||||
GenBank mRNA | AF030091.3 | - | 1351 | |||
BC085686.1 | - | 1405 | ||||
BC128699.1 | - | 1445 | ||||
BC161817.1 | - | 1297 | 9022416 | |||
FQ213168.1 | - | 8 | 775 |
