siRNA Details
| Gene Symbol: Akap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053665.1 | NP_446117.1 | 1 | 409 | ||
| XM_006247067.2 | XP_006247129.1 | 2 | 582 | |||
| XM_006247068.1 | XP_006247130.1 | 2 | 578 | |||
| XM_008767930.1 | XP_008766152.1 | 2 | 637 | |||
| GenBank mRNA | AF068202.1 | - | 1 | 409 | ||
| AF092523.1 | - | 1 | 409 | |||
| BC081753.1 | - | 2 | 558 | |||
| BC097298.1 | - | 2 | 570 | |||
| BC129090.1 | - | 2 | 549 | |||

Akap1
Genomic Map