siRNA Details
| Gene Symbol: Mpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053668.1 | NP_446120.1 | 6 | 554 | ||
| XM_008767931.1 | XP_008766153.1 | 6 | 530 | |||
| XM_008767932.1 | XP_008766154.1 | 6 | 654 | |||
| GenBank mRNA | AF087697.1 | - | 4 | 406 | ||
| FQ211573.1 | - | 6 | 680 | |||

Mpp3
Genomic Map