siRNA Details
| Gene Symbol: Mtnr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053676.2 | NP_446128.1 | 2 | 443 | ||
| GenBank mRNA | AB377274.1 | - | 2 | 444 | ||
| AF130341.1 | - | 2 | 219 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtnr1a| Gene Symbol: Mtnr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053676.2 | NP_446128.1 | 2 | 443 | ||
| GenBank mRNA | AB377274.1 | - | 2 | 444 | ||
| AF130341.1 | - | 2 | 219 | |||
Set Membership: |
|