siRNA Details
| Gene Symbol: Nox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053683.1 | NP_446135.1 | 10 | 1360 | ||
| GenBank mRNA | AF152963.1 | - | 10 | 1360 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nox1| Gene Symbol: Nox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053683.1 | NP_446135.1 | 10 | 1360 | ||
| GenBank mRNA | AF152963.1 | - | 10 | 1360 | ||