siRNA Details
| Gene Symbol: Hcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053685.1 | NP_446137.1 | 5 | 1158 | ||
| GenBank mRNA | AF247452.1 | - | 5 | 1158 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcn3| Gene Symbol: Hcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053685.1 | NP_446137.1 | 5 | 1158 | ||
| GenBank mRNA | AF247452.1 | - | 5 | 1158 | ||