siRNA Details
| Gene Symbol: Trpv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053686.1 | NP_446138.1 | 13 | 1916 | ||
| XM_006236375.2 | XP_006236437.1 | 13 | 1532 | |||
| GenBank mRNA | AF160798.1 | - | 13 | 1916 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpv6| Gene Symbol: Trpv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053686.1 | NP_446138.1 | 13 | 1916 | ||
| XM_006236375.2 | XP_006236437.1 | 13 | 1532 | |||
| GenBank mRNA | AF160798.1 | - | 13 | 1916 | ||