siRNA Details
| Gene Symbol: Cdk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191861.1 | NP_001178790.1 | 2 | 305 | ||
| XM_006236019.2 | XP_006236081.1 | 3 | 769 | |||
| XM_006236020.2 | XP_006236082.1 | 3 | 703 | |||
| XM_006236021.1 | XP_006236083.1 | 3 | 645 | |||
| XM_008762679.1 | XP_008760901.1 | 3 | 620 | |||
| GenBank mRNA | AF352168.1 | - | 2 | 233 | ||

Cdk6
Genomic Map