siRNA Details
| Gene Symbol: Dag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053697.1 | NP_446149.1 | 2 | 1426 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dag1| Gene Symbol: Dag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053697.1 | NP_446149.1 | 2 | 1426 | ||