siRNA Details
| Gene Symbol: Cacna1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053701.1 | NP_446153.1 | 13 | 1622 | ||
| XM_006256698.2 | XP_006256760.1 | 9 | 999 | |||
| GenBank mRNA | AF365975.1 | - | 13 | 1622 | ||
| DQ393415.1 | - | 13 | 1626 | |||

Cacna1f
Genomic Map