siRNA Details
| Gene Symbol: Cacna1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053701.1 | NP_446153.1 | 4 | 518 | ||
| GenBank mRNA | AF365975.1 | - | 4 | 518 | ||
| DQ393415.1 | - | 4 | 522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna1f| Gene Symbol: Cacna1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053701.1 | NP_446153.1 | 4 | 518 | ||
| GenBank mRNA | AF365975.1 | - | 4 | 518 | ||
| DQ393415.1 | - | 4 | 522 | |||