siRNA Details
| Gene Symbol: Map2k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053703.3 | NP_446155.1 | 8 | 656 | ||
| GenBank mRNA | AF369384.1 | - | 8 | 656 | ||
| BC087004.1 | - | 8 | 756 | 7190965 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map2k6| Gene Symbol: Map2k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053703.3 | NP_446155.1 | 8 | 656 | ||
| GenBank mRNA | AF369384.1 | - | 8 | 656 | ||
| BC087004.1 | - | 8 | 756 | 7190965 | ||