siRNA Details
| Gene Symbol: Pax3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053710.1 | NP_446162.1 | 4 | 852 | ||
| XM_006245131.2 | XP_006245193.1 | 4 | 906 | |||
| XM_008767181.1 | XP_008765403.1 | 4 | 910 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pax3| Gene Symbol: Pax3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053710.1 | NP_446162.1 | 4 | 852 | ||
| XM_006245131.2 | XP_006245193.1 | 4 | 906 | |||
| XM_008767181.1 | XP_008765403.1 | 4 | 910 | |||